VWR® ARISTAR®, Scandium Standard Solution, 10000 mg/L Sc in dil. Nitric Acid (from Sc2O3), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455972UEA
852
EUR
455972U
455974W
VWR® ARISTAR®, Scandium Standard Solution, 10000 mg/L Sc in dil. Nitric Acid (from Sc2O3), Standard for ICP
Standaarden
Kationische standaarden
Scandium standaarden
Sc₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.