VWR® ARISTAR®, Selenium Standard Solution, 1000 mg/L Se in dil. Nitric Acid (from Se), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455982WEA 96 EUR
455982W 455984B
VWR® ARISTAR®, Selenium Standard Solution, 1000 mg/L Se in dil. Nitric Acid (from Se), Standard for ICP
Standaarden Kationische standaarden Selenium standaarden
Se in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.