VWR® ARISTAR®, Silicon Standard Solution, 1000 mg/L Si in Water with Hydrofluoric Acid (max. 1%) (from (NH4)2SiF6), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456002GEA 103.8 EUR
456002G 456004Y
VWR® ARISTAR®, Silicon Standard Solution, 1000 mg/L Si in Water with Hydrofluoric Acid (max. 1%) (from (NH4)2SiF6), Standard for ICP
Standaarden Kationische standaarden Silicium standaarden
(NH₄)₂ SiF₆ in H₂O tr. HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.
Order Now
Specificaties
Environmentally Preferable