Zone samplers for powders and granules
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1610A-1000 1610A-1500 1610A-2000 1610A-2500 1610A-600
300-0443EA 272 EUR
300-0443 300-0444 300-0445 300-0446 300-0447
Zone samplers for powders and granules
Monsternemers Monsternemers voor vaste stoffen
An ideal tool for taking representative composite samples from containers, big bags or silos.
  • Aluminium construction
  • Takes multiple samples along its length
  • Easy to operate
The zone sampler is ideal for taking samples of powders and granules at multiple locations. The lightweight sampler is quick and easy to use.
The robust double tube design keeps material out of the sampler until it is opened.