VWR® ARISTAR®, Strontium Standard Solution, 1000 mg/L Sr in dil. Nitric Acid (from Sr(NO₃)₂), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456062SEA
162
EUR
456062S
456064U
VWR® ARISTAR®, Strontium Standard Solution, 1000 mg/L Sr in dil. Nitric Acid (from Sr(NO₃)₂), Standard for ICP
Standaarden
Kationische standaarden
Strontium standaarden
Sr(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.