VWR® ARISTAR®, Tellurium Standard Solution, 1000 mg/L Te in 20% Hydrochloric Acid (from Te), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456122NEA
157.2
EUR
456122N
456124P
VWR® ARISTAR®, Tellurium Standard Solution, 1000 mg/L Te in 20% Hydrochloric Acid (from Te), Standard for ICP
Standaarden
Kationische standaarden
Tellurium standaarden
Te in HCl 20%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.