Terbium standard solution, 1000 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13860.AD
13860.AE
13860.AP
13860.ADEA
69.3
EUR
13860.AD
13860.AE
13860.AP
Terbium standard solution, 1000 mg/L Tb in 5% nitric acid (from Tb₄O₇), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Terbium standaarden
Molecular formula: Tb₄O₇ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011256
Certificaten: Traceable to NIST.