VWR® ARISTAR®, Vanadium Standard Solution, 1000 mg/L V in dil. Nitric Acid (from V2O5), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456322TEA
94.8
EUR
456322T
456324V
VWR® ARISTAR®, Vanadium Standard Solution, 1000 mg/L V in dil. Nitric Acid (from V2O5), Standard for ICP
Standaarden
Kationische standaarden
Vanadium standaarden
V₂O₅ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.