VWR® AVS TITRINORM, Vanadium Standard Solution, 1000 mg/L V in 2% sulphuric Acid, Standard for AAS
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
86721.260EA
25.69
EUR
86721.260
86721.180
VWR® AVS TITRINORM, Vanadium Standard Solution, 1000 mg/L V in 2% sulphuric Acid, Standard for AAS
Standaarden
Kationische standaarden
Vanadium standaarden
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%