VWR® ARISTAR®, Vanadium Standard Solution, 10000 mg/L V in dil. Nitric Acid (from V2O5), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456332VEA 126 EUR
456332V 456334A
VWR® ARISTAR®, Vanadium Standard Solution, 10000 mg/L V in dil. Nitric Acid (from V2O5), Standard for ICP
Standaarden Kationische standaarden Vanadium standaarden
V₂O₅ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.