VWR® ARISTAR®, Tungsten Standard Solution, 1000 mg/L W in Water (from W), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
457182GEA 256 EUR
457182G 457184Y
VWR® ARISTAR®, Tungsten Standard Solution, 1000 mg/L W in Water (from W), Standard for ICP
Standaarden Kationische standaarden Wolfraam standaarden
W in H₂O tr.NH₄OH.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.