Ytterbium standard solution, 10000 mg/L Yb in 5% nitric acid (from Yb₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
14439.AE
14439.AP
14439.AEEA
155
EUR
14439.AE
14439.AP
Ytterbium standard solution, 10000 mg/L Yb in 5% nitric acid (from Yb₂O₃), standard for ICP, Specpure®
Standaarden
Kationische standaarden
Ytterbium standaarden
Molecular formula: Yb₂O₃ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011286
Certificaten: Traceable to NIST.