Ytterbium standard solution, 1000 mg/L Yb in 5% nitric acid (from Yb₂O₃), standard for ICP, Specpure®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
13819.AD 13819.AE 13819.AP
13819.ADEA 68.3 EUR
13819.AD 13819.AE 13819.AP
Ytterbium standard solution, 1000 mg/L Yb in 5% nitric acid (from Yb₂O₃), standard for ICP, Specpure®
Standaarden Kationische standaarden Ytterbium standaarden
Molecular formula: Yb₂O₃ in 5% HNO₃
Physical form: liquid
UN No.: 3264
MDL No.: MFCD00011286
Certificaten: Traceable to NIST.