VWR®, Yttrium standard solution, 1000 mg/L Y in dil. nitric acid (from Y₂O₃) ARISTAR® standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456362EEA 160 EUR
456362E 456364G
VWR®, Yttrium standard solution, 1000 mg/L Y in dil. nitric acid (from Y₂O₃) ARISTAR® standard for ICP
Standaarden Kationische standaarden Yttrium standaarden
Y₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.