Yttrium standard solution, 10.000 mg/l Y in dil. nitric acid (from Y₂O₃) ARISTAR® standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456372GEA 319 EUR
456372G 456374Y
Yttrium standard solution, 10.000 mg/l Y in dil. nitric acid (from Y₂O₃) ARISTAR® standard for ICP
Standaarden Kationische standaarden Yttrium standaarden
Y₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Certificaten: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Leveringsinformatie: Supplied with certificate of analysis.