VWR® ARISTAR®, Zinc Standard Solution, 10000 mg/L Zn in dil. Nitric Acid (from Zn), Standard for ICP
Leverancier: VWR Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456392KEA
309
EUR
456392K
456394M
VWR® ARISTAR®, Zinc Standard Solution, 10000 mg/L Zn in dil. Nitric Acid (from Zn), Standard for ICP
Standaarden
Kationische standaarden
Zink standaarden
Zn in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.