VWR® ARISTAR®, Tin Standard Solution, 1000 mg/L Sn in dil. Nitric Acid with Hydrofluoric Acid (max. 1%) (from Sn), Standard for ICP
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
456222QEA 160 EUR
456222Q 456224S
VWR® ARISTAR®, Tin Standard Solution, 1000 mg/L Sn in dil. Nitric Acid with Hydrofluoric Acid (max. 1%) (from Sn), Standard for ICP
Standaarden Kationische standaarden Tin standaarden
Sn in HNO₃ tr. HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.