Potassium iodate-iodide (0,713 g/L KIO₃) 0.02 N in aqueous solution stabilised volumetric solution
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
160274XEA 55.7 EUR
160274X 160276Q
Potassium iodate-iodide (0,713 g/L KIO₃) 0.02 N in aqueous solution stabilised volumetric solution
Potassium iodate-iodide
Standard solution for the determination of sulphite in boiler water.
Order Now
Specification Test Results
Assay (Potassium iodate) 0.7126 - 0.7140 g/l