Kaliumdichromaat standardised solution 0.25 N in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
35587.K2 35587.AP
35587.K2EA 48.7 EUR
35587.K2 35587.AP
Kaliumdichromaat standardised solution 0.25 N in aqueous solution
Kaliumdichromaat
CAS No.: 7778-50-9 Molecular Formula: Cr2K2O7 Formula Weight: 294.18 Physical Form: Liquid Appearance: Orange Odour: Odourless UN No.: 3287 MDL No.: MFCD00011367
Formule: K₂Cr₂O₇
MW: 294.18 g/mol
Kookpunt: 500
Smeltpunt: 398
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011367
CAS nummer: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1 III
Merck Index: 12,07790