Calcium acetate monohydrate ≥99% ACS
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
|
Synoniemen:
Calcium di(acetate) monohydrate, Calcium acetate hydrate, Acetic acid calcium salt monohydrate
13446.30
13446.A1
13446.30EA
37.2
EUR
13446.30
13446.A1
Calcium acetate monohydrate ≥99% ACS
Calcium acetate monohydrate
Purity: 99.0% min
CAS No.: 5743-26-0
Molecular Formula: C4H6CaO4.H2O
Formula Weight: 176.18 (158.17anhy)
Physical Form: Powder
Density: 1.5
Appearance: White
Melting Point: dec.
Beilstein Reference: 3692527
Merck Reference: 14,1646
MDL No.: MFCD00150019
Formule:
(H₃CCOO)₂Ca·1H₂O MW: 176.18 g/mol Smeltpunt: Dec. Dichtheid: 1.5 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150019 CAS nummer: 5743-26-0 EINECS: 200-540-9 Merck Index: 14,1646 |