Tintetrachloride 1 mol/l dichloromethane, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Stannic chloride, Tin tetrachloride
378201000 378208000
378201000.EA 62.9 EUR
378201000. 378208000.
Tintetrachloride 1 mol/l dichloromethane, AcroSeal®
Tintetrachloride
Formule: Cl₄Sn
MW: 260.52 g/mol
Kookpunt: 114
Smeltpunt: -33
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011242
CAS nummer: 7646-78-8
EINECS: 231-588-9
UN: 2922
Merck Index: 12,08929
Order Now
Specification Test Results
Appearance Clear colorless Solution
Titration Argentometric 0.95 to 1.10 M