Calcium acetate xhydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Acetic acid calcium salt hydrate, Calcium di(acetate) hydrate, Calcium acetate hydrate
P011.1 P011.2
ROTHP011.1EA 66.5 EUR
ROTHP011.1 ROTHP011.2
Calcium acetate xhydrate
Calcium acetate xhydrate
Formule: (H₃CCOO)₂Ca·nH₂O
MW: 176.18 g/mol
Dichtheid: 1.5 g/cm³ (20 °C)
MDL Nummer: MFCD00150019
CAS nummer: 114460-21-8
EINECS: 200-540-9