Volumetric measuring scoops, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8286A-1000S 8286A-500S 8286A-50S 8286A-500 8286A-50 8286A-25S 8286A-250S 8286A-250 8286A-25 8286A-150S 8286A-150 8286A-100S 8286A-1000 8286A-100
300-0432EA 333 EUR
300-0432 300-0666 300-0568 300-0569 300-0570 300-0571 300-0572 300-0573 300-0574 300-0575 300-0576 300-0577 300-0578 300-0579
Volumetric measuring scoops, SteriWare®
Scheppen
These volumetric scoops are ideal for measuring set amounts from 25 ml to 1 L. They are ideal for use in laboratory to production. The flat base means they are sturdy and easy to use when put down.
  • Robust PS construction
  • Available in wide range of sizes from 25 ml to 1000 ml
  • Produced in a cleanroom
  • Ergonomic handle and heavy duty design
  • Easy to use
Both non sterile and sterile available. Sterile scoops conform to SAL 10⁻⁶. Sterilisation is done by gamma irradiation.
If measuring powder after scooping remove the excess powder to give an accurate volume.
Certificaten: Conform to the latest standards including FDA 177.1640, EU 10/2011, EC 1935/2004 and are BSE-/TSE-free.
Verpakking: Individually bagged.