Sodium diethyldithiocarbamate trihydrate, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (Diethyldithiocarbamato-S,S')sodium trihydrate, Diethyldithiocarbamic acid sodium salt trihydrate, N,N-Diethyldithiocarbamic acid sodium salt trihydrate
71480-100G 71480-500G
71480-100GEA 75.5 EUR
71480-100G 71480-500G
Sodium diethyldithiocarbamate trihydrate, Supelco®
Sodium diethyldithiocarbamate trihydrate
Formule: (C₂H₅)₂NCSSNa·3H₂O
MW: 225.31 g/mol
Smeltpunt: 95…98.5 °C
Dichtheid: 1.1 g/cm³ (20 °C)
MDL Nummer: MFCD00150617
CAS nummer: 20624-25-3
EINECS: 205-710-6
UN: 3077
ADR: 9 III
Merck Index: 12,03443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98fc677309a4c79e2381c3545fa9a760182d4bc0&vid=98fc677309a4c79e2381c3545fa9a760182d4bc0&prid=web&clid=SAPDAM