Phosphotungstic acid hydrate, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Phosphotungstic acid hydrate, Wolframophosphoric acid hydrate, Dodecawolframophosphoric acid hydrate, 12-Tungstophosphate hydrate, Tungstophosphoric Acid Hydrate
M-Clarity™ quality level = MQ200
1.00582.0100 1.00582.9025
1.00582.0100EA 166 EUR
1.00582.0100 1.00582.9025
Phosphotungstic acid hydrate, Supelco®
Phosphotungstic acid hydrate
Formule: H₃[P(W₃O₁₀)₄]·nH₂O
Smeltpunt: 107 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011342
CAS nummer: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 13,07450
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 107 °C
Molar Mass 2880.17 g/mol (anhydrous)
Bulk Density 960 kg/m3
pH 1.3 (20 g/l, H2O, 20 °C)