Hexachloroplatinic(IV) acid hexahydrate ≥99.95% (metals basis) Pt ≥ 37.5%, Premion® ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate
36259.03 36259.06 36259.14
36259.03EA 127 EUR
36259.03 36259.06 36259.14
Hexachloroplatinic(IV) acid hexahydrate ≥99.95% (metals basis) Pt ≥ 37.5%, Premion® ACS
Hexachloroplatinic(IV) acid hexahydrate
Purity: 99.95% CAS No.: 18497-13-7 Molecular Formula: H2Cl6Pt.6H2O Formula Weight: 517.91 (409.82anhy) Physical Form: Lump Density: 2.430 Appearance: Red-brown Odour: Odourless Melting Point: 60° dec. Merck Reference: 14,7526 UN No.: 2507 MDL No.: MFCD00149910
Formule: H₂PtCl₆·6H₂O
MW: 517.9 g/mol
Smeltpunt: 60 °C (ontbinding)
Dichtheid: 2.430
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149910
CAS nummer: 18497-13-7
EINECS: 241-010-7
UN: 2507
ADR: 8 III
Merck Index: 14,7526