Sodium hypophosphite monohydrate
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium phosphinate monohydrate
14104.30 14104.A1
14104.30EA 36.6 EUR
14104.30 14104.A1
Sodium hypophosphite monohydrate
Sodium hypophosphite monohydrate
Formule: NaH₂PO₂·H₂O
MW: 105.99 g/mol
Smeltpunt: 90
Dichtheid: 0.8 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06252379
CAS nummer: 10039-56-2
EINECS: 231-669-9
Merck Index: 14,8629