Disodium hydrogen phosphate dodecahydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate
T106.1 T106.2
ROTHT106.1EA 75.9 EUR
ROTHT106.1 ROTHT106.2
Disodium hydrogen phosphate dodecahydrate
Disodium hydrogen phosphate dodecahydrate
Formule: Na₂HPO₄·12H₂O
MW: 358.14 g/mol
Smeltpunt: 35
Dichtheid: 1.52 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149181
CAS nummer: 10039-32-4
EINECS: 231-448-7
Merck Index: 13,08733