Disodium hydrogen phosphate dihydrate
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: sec-Sodium phosphate, Sodium monohydrogen phosphate dihydrate, Sodium hydrogenphosphate dihydrate, di-Sodium hydrogen orthophosphate dihydrate, Sodium phosphate dibasic dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate, Sorensen's salt
39783.01 39783.02
SERA39783.01EA 0 EUR
SERA39783.01 SERA39783.02
Disodium hydrogen phosphate dihydrate
Disodium hydrogen phosphate dihydrate
Formule: Na₂HPO₄·2H₂O
MW: 177.99 g/mol
Smeltpunt: 92.5
Dichtheid: 2.1 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149182
CAS nummer: 10028-24-7
EINECS: 231-448-7