Disodium hydrogen phosphate heptahydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium phosphate dibasic heptahydrate, Sodium hydrogen phosphate heptahydrate, Sodium monohydrogen phosphate heptahydrate
X987.1 X987.2
ROTHX987.1EA 27.5 EUR
ROTHX987.1 ROTHX987.2
Disodium hydrogen phosphate heptahydrate
Disodium hydrogen phosphate heptahydrate
Formule: HNa₂O₄P·7H₂O
MW: 268.07 g/mol
Smeltpunt: 48
Dichtheid: 1.7 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149180
CAS nummer: 7782-85-6
EINECS: 231-448-7