L(-)-Cystine

Leverancier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

17880.01 17880.02 17880.03
SERA17880.01EA 0 EUR
SERA17880.01 SERA17880.02 SERA17880.03
L(-)-Cystine
L(-)-Cystine
Formule: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
Kookpunt: 428 °C (1013 hPa)
Smeltpunt: 261…262 °C
Dichtheid: 1,677 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064228
CAS nummer: 56-89-3
EINECS: 200-296-3
Merck Index: 13,02811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR