Zinc perchlorate hexahydrate ≥99.997% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
44315.09
44315.18
44315.09EA
88
EUR
44315.09
44315.18
Zinc perchlorate hexahydrate ≥99.997% (metals basis)
Zinc perchlorate hexahydrate
Purity: 99.997%
CAS No.: 10025-64-6
Molecular Formula: Cl2O8Zn.6H2O
Formula Weight: 372.37 (264.28anhy)
Physical Form: Crystalline
Density: 2.25
Refractive Index: 1.50
Appearance: White
Odour: Odourless
Melting Point: 106°
Merck Reference: 14,10148
UN No.: 1481
MDL No.: MFCD00150370
Formule:
Cl₂O₈Zn·6H₂O Smeltpunt: 106 °C Dichtheid: 2,25 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150370 CAS nummer: 10025-64-6 EINECS: 237-122-0 UN: 1481 ADR: 5.1,II Merck Index: 14,10148 |