Cobalt(II) bromide hydrate
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Cobalt dibromide hydrate, Cobaltous bromide hydrate
40225.18
40225.30
40225.18EA
87.5
EUR
40225.18
40225.30
Cobalt(II) bromide hydrate
Cobalt(II) bromide hydrate
CAS No.: 85017-77-2
Molecular Formula: Br2Co.xH2O
Formula Weight: 218.75(anhy)
Physical Form: Crystalline Granular
Density: 2.46
Sensitivity: Hygroscopic
Appearance: Red-purple
Odour: Odourless
Melting Point: 47-48° dec.
Merck Reference: 14,2435
MDL No.: MFCD00661152
Hygrometers, catalysts
Formule:
Br₂Co·H₂O Dichtheid: 2,46 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00150208 CAS nummer: 85017-77-2 EINECS: 232-166-7 Merck Index: 14,2435 |