Nickel(II) perchlorate hexahydrate, Reagent Grade
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
CAS No.: 13520-61-1
Molecular Formula: Ni(ClO4)2.6H2O
Formula Weight: 365.70 (257.59anhy)
Physical Form: Damp crystalline solid
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: Green
Odour: Odourless
Melting Point: 140°
Solubility: Very soluble in water. Soluble in many organic solvents
UN No.: 1481
MDL No.: MFCD00149804
Formule:
Cl₂NiO₈·6H₂O Smeltpunt: 140 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149804 CAS nummer: 13520-61-1 EINECS: 237-124-1 UN: 1481 ADR: 5.1,II |