Nickel(II) perchlorate hydrate ≥99.998% (metals basis)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
44316.09
44316.18
44316.09EA
69
EUR
44316.09
44316.18
Nickel(II) perchlorate hydrate ≥99.998% (metals basis)
Nickel(II) perchlorate hydrate
Purity: 99.998%
CAS No.: 13637-71-3
Molecular Formula: Cl2NiO8.xH2O
Formula Weight: 257.59(anhy)
Physical Form: Crystalline
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: Green
Odour: Odourless
Melting Point: 140°
UN No.: 1481
MDL No.: MFCD00149804
Formule:
Cl₂NiO₈·xH₂O Smeltpunt: 140 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149804 CAS nummer: 13637-71-3 EINECS: 237-124-1 UN: 1481 ADR: 5.1,II |