1-Cetylpyridinium chloride monohydrate 98
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate
A13499.14
A13499.22
A13499.36
A13499.14EA
23.1
EUR
A13499.14
A13499.22
A13499.36
1-Cetylpyridinium chloride monohydrate 98
1-Cetylpyridinium chloride monohydrate
Purity: 98%
CAS No.: 6004-24-6
Molecular Formula: C21H38ClN.H2O
Formula Weight: 358.02 (339.99anhy)
Melting Point: 83-86°
Solubility: Soluble in water, alcohol, chloroform
Beilstein Reference: 3578606
Merck Reference: 14,2032
UN No.: 2811
MDL No.: MFCD00149977
Formule:
C₂₁H₃₈ClN·1H₂O MW: 358.01 g/mol Smeltpunt: 83…86 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149977 CAS nummer: 6004-24-6 EINECS: 204-593-9 UN: 2811 ADR: 6.1 II Merck Index: 13,02041 |