meta-Phosphoric acid stabilised
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Metaphosphoric acid
219221000 219225000
ACRO219221000EA 66.8 EUR
ACRO219221000 ACRO219225000
meta-Phosphoric acid stabilised
meta-Phosphoric acid
Formule: (HPO₃)n
MW: 79.98 g/mol
Kookpunt: 260
Smeltpunt: 21
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011351
CAS nummer: 37267-86-0
EINECS: 253-433-4
UN: 3260
ADR: 8 III
Order Now
Specification Test Results
Appearance Colorless to white Lumps
Stabilizer 54 to 60 % NaPO3
Titration with NaOH 40 to 46 %
Chloride (Cl) ≤20 ppm
Arsenic (As) ≤5 ppm
Iron (Fe) ≤100 ppm
Heavy metals (as Pb) ≤50 ppm