Carbazochrome ≥99.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A0176-25G A0176-5G
TCIAA0176-25GEA 162 EUR
TCIAA0176-25G TCIAA0176-5G
Carbazochrome ≥99.0% (by HPLC)
Carbazochrome
Formule: C₁₀H₁₂N₄O₃
MDL Nummer: MFCD00059705
CAS nummer: 69-81-8
EINECS: 200-717-0
Merck Index: 14,00172

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 99.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR