Acetaldehyde semicarbazone ≥98.0% (by titrimetric analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Acetaldehyde Semicarbazone
A0193-25G
A0193-5G
TCIAA0193-25GEA
190
EUR
TCIAA0193-25G
TCIAA0193-5G
Acetaldehyde semicarbazone ≥98.0% (by titrimetric analysis)
Acetaldehyde semicarbazone
Formule:
C₃H₇N₃O MW: 101.11 g/mol |
MDL Nummer:
MFCD00059191 CAS nummer: 591-86-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Potassium iodate Method) | min. 98.0 % |
Melting point | 163.0 to 166.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...