Sodium-4-aminobenzoate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
A0276-25G
A0276-500G
TCIAA0276-25GEA
26.4
EUR
TCIAA0276-25G
TCIAA0276-500G
Sodium-4-aminobenzoate ≥98.0% (by HPLC, titration analysis)
Sodium-4-aminobenzoate
Formule:
C₇H₇NO₂ |
MDL Nummer:
MFCD00064395 CAS nummer: 555-06-6 EINECS: 209-080-3 |
Specification Test Results
Appearance | White to Light yellow to Light red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...