Amylopectin hydrate (from waxy corn) amylose-free
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A0456-500G A0456-25G
TCIAA0456-500GEA 44 EUR
TCIAA0456-500G TCIAA0456-25G
Amylopectin hydrate (from waxy corn) amylose-free
Amylopectin hydrate
Formule: (C₆H₁₀O₅)n·xH₂O
MDL Nummer: MFCD00130510
CAS nummer: 9037-22-3
EINECS: 232-911-6
Merck Index: 14,00481
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Total Nitrogen max. 0.06 %
Ash Content max. 0.1 %
Drying loss max. 14.0 %