Trimethyl-trans-aconitate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: trans-Aconitic Acid Trimethyl Ester

A0725-25G A0725-5G
TCIAA0725-25GEA 294 EUR
TCIAA0725-25G TCIAA0725-5G
Trimethyl-trans-aconitate ≥97.0% (by GC)
Trimethyl-trans-aconitate
Formule: C₉H₁₂O₆
Kookpunt: 160°C
MDL Nummer: MFCD00059304
CAS nummer: 4271-99-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR