Dimethyl adipimidate dihydrochloride ≥98.0% (by titrimetric analysis) cross-linking agent, for protein research

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Adipimidic Acid Dimethyl Ester Dihydrochloride

A0806-25G A0806-5G
TCIAA0806-25GEA 146 EUR
TCIAA0806-25G TCIAA0806-5G
Dimethyl adipimidate dihydrochloride ≥98.0% (by titrimetric analysis) cross-linking agent, for protein research
Dimethyl adipimidate dihydrochloride
Formule: C₈H₁₆N₂O₂
MW: 245.14 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00012573
CAS nummer: 14620-72-5
EINECS: 238-660-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR