Acetyl sulphide ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Diacetyl Sulphide
A0976-25G A0976-5G
TCIAA0976-25GEA 541 EUR
TCIAA0976-25G TCIAA0976-5G
Acetyl sulphide ≥95.0% (by GC)
Acetyl sulphide
Formule: C₄H₆O₂S
MW: 118.15 g/mol
Kookpunt: 120°C
Vlampunt: 27°C
MDL Nummer: MFCD00008753
CAS nummer: 3232-39-1
EINECS: 221-774-8
UN: 1993
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19da4c399d2cbe76e3ebcda44b149aa113ed1b07&vid=19da4c399d2cbe76e3ebcda44b149aa113ed1b07&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %