Stearyl acrylate ≥97.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing


Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

A1011-25G A1011-400G
TCIAA1011-25GEA 17.6 EUR
TCIAA1011-25G TCIAA1011-400G
Stearyl acrylate ≥97.0% (by GC) stabilised
Stearyl acrylate
Formule: C₂₁H₄₀O₂
MDL Nummer: MFCD00015091
CAS nummer: 4813-57-4
EINECS: 225-383-3

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %
Solubility in hot Methanol very faint turbidity

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR