Methyl dichloromethoxyacetate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

A1046-25G A1046-5G
TCIAA1046-25GEA 236 EUR
TCIAA1046-25G TCIAA1046-5G
Methyl dichloromethoxyacetate ≥95.0% (by GC)
Methyl dichloromethoxyacetate
Formule: C₄H₆Cl₂O₃
Kookpunt: 181°C
Vlampunt: 91°C
MDL Nummer: MFCD00012202
CAS nummer: 17640-25-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bccf2e2b76d2896e2ce17cc69fee670319474d7&vid=9bccf2e2b76d2896e2ce17cc69fee670319474d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR