(-)-O-Acetyl-L-malic anhydride ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (S)-2-Acetoxysuccinic Anhydride, (S)-(-)-2-Acetoxysuccinic Anhydride

A1101-25G A1101-5G
TCIAA1101-25GEA 237 EUR
TCIAA1101-25G TCIAA1101-5G
(-)-O-Acetyl-L-malic anhydride ≥97.0% (by titrimetric analysis)
(-)-O-Acetyl-L-malic anhydride
Formule: C₆H₆O₅
MDL Nummer: MFCD00082708
CAS nummer: 59025-03-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Methanolysis Method) min. 97.0 %
Melting point 54.0 to 59.0 deg-C
Specific rotation [a]20/D -24.0 to -27.0 deg (C=5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR