3-Ethynylaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

A1122-25G A1122-5G
TCIAA1122-25GEA 90.2 EUR
TCIAA1122-25G TCIAA1122-5G
3-Ethynylaniline ≥98.0% (by GC)
3-Ethynylaniline
Formule: C₈H₇N
Kookpunt: 120°C
MDL Nummer: MFCD00014779
CAS nummer: 54060-30-9
EINECS: 258-944-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f24cdde25e64071ea0c8bcdd9c5212696dbf6b9&vid=3f24cdde25e64071ea0c8bcdd9c5212696dbf6b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR