2-Aminoethyl isopropyl ether ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

Synoniemen: 2-Isopropoxyethylamine, 2-Isopropoxyethanamine

A1296-25ML A1296-5ML
TCIAA1296-25MLEA 613 EUR
TCIAA1296-25ML TCIAA1296-5ML
2-Aminoethyl isopropyl ether ≥98.0% (by GC, titration analysis)
2-Aminoethyl isopropyl ether
Formule: C₅H₁₃NO
Kookpunt: 120°C
MDL Nummer: MFCD00191603
CAS nummer: 81731-43-3
UN: 2733
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR