2-Phenoxyethyl acrylate ≥93.0% (by GC) stabilised

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Acrylic Acid 2-Phenoxyethyl Ester, Ethylene Glycol Monophenyl Ether Acrylate

Stabilisatie: Stabilised with 4-methoxyphenol (MEHQ)

A1400-25G A1400-500G
TCIAA1400-25GEA 17.6 EUR
TCIAA1400-25G TCIAA1400-500G
2-Phenoxyethyl acrylate ≥93.0% (by GC) stabilised
2-Phenoxyethyl acrylate
Formule: C₁₁H₁₂O₃
Vlampunt: 141°C
MDL Nummer: MFCD00053696
CAS nummer: 48145-04-6
EINECS: 256-360-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 93.0 %
Acid Value max. 1.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR